Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ama-1
Entrez Gene ID
177190
Gene Name
DNA-directed RNA polymerase II subunit RPB1
Gene Aliases
CELE_F36A4.7
Chromosome Location
Chr.IV: 4248020 - 4258439 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068122.8 NP_500523.4 9 - 10 3604 71

View Full Product Details