Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-52
Entrez Gene ID
189147
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_W03B1.8
Chromosome Location
Chr.IV: 4342262 - 4346612 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068145.3 NP_500546.2 6 - 7 1089 142

View Full Product Details