Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sar-1
Entrez Gene ID
177217
Gene Name
GTP-binding protein SAR1
Gene Aliases
CELE_ZK180.4
Chromosome Location
Chr.IV: 4511892 - 4513980 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068181.5 NP_500582.1 3 - 4 225 96

View Full Product Details