Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eor-1
Entrez Gene ID
177249
Gene Name
protein-coding
Gene Aliases
CELE_R11E3.6
Chromosome Location
Chr.IV: 4793194 - 4799187 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380207.2 NP_001367921.1 9 - 10 1525 103
GenBank mRNA AF502568.1 - 10 - 11 1633 103
AF519108.1 - 9 - 10 1514 103

View Full Product Details