Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eor-1
Entrez Gene ID
177249
Gene Name
protein-coding
Gene Aliases
CELE_R11E3.6
Chromosome Location
Chr.IV: 4793194 - 4799187 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380207.2 NP_001367921.1 3 - 4 480 154
GenBank mRNA AF502568.1 - 4 - 5 588 154
AF519108.1 - 3 - 4 469 154

View Full Product Details