Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sand-1
Entrez Gene ID
177318
Gene Name
SAND endocytosis protein family
Gene Aliases
CELE_F41H10.11
Chromosome Location
Chr.IV: 5366867 - 5368865 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068390.9 NP_500791.2 3 - 4 691 111

View Full Product Details