Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elo-5
Entrez Gene ID
177320
Gene Name
Elongation of very long chain fatty acids protein 5
Gene Aliases
CELE_F41H10.7
Chromosome Location
Chr.IV: 5374598 - 5379757 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380227.2 NP_001368393.1 4 - 5 884 136

View Full Product Details