Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-33
Entrez Gene ID
177361
Gene Name
protein-coding
Gene Aliases
CELE_H32C10.2
Chromosome Location
Chr.IV: 5930645 - 5933326 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380247.1 NP_001368368.1 1 - 2 188 86
NM_068489.5 NP_500890.1 3 - 4 428 86

View Full Product Details