Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-20
Entrez Gene ID
187130
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_K08B4.4
Chromosome Location
Chr.IV: 6080895 - 6083600 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068512.3 NP_500913.1 3 - 4 453 98

View Full Product Details