The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_068573.9 | NP_500974.2 | 11 - 12 | 2963 | 78 |
GenBank mRNA | EF472000.1 | - | 11 - 12 | 2977 | 78 |