Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tag-30
Entrez Gene ID
178116
Gene Name
BTB domain-containing protein
Gene Aliases
CELE_F38H4.7
Chromosome Location
Chr.IV: 11850928 - 11856379 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380473.1 NP_001366838.1 7 - 8 1215 69

View Full Product Details