Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mig-32
Entrez Gene ID
178150
Gene Name
protein-coding
Gene Aliases
CELE_F11A10.3
Chromosome Location
Chr.IV: 12089286 - 12092213 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268691.2 NP_001255620.1 2 - 3 1477 73

View Full Product Details