Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-14
Entrez Gene ID
183397
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_C42C1.1
Chromosome Location
Chr.IV: 12259793 - 12261453 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069927.4 NP_502328.2 2 - 3 323 118

View Full Product Details