Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ham-1
Entrez Gene ID
178205
Gene Name
Stork_head domain-containing protein
Gene Aliases
CELE_F53B2.6
Chromosome Location
Chr.IV: 12542034 - 12545737 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070000.7 NP_502401.1 1 - 2 281 109

View Full Product Details