Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pin-2
Entrez Gene ID
178234
Gene Name
protein-coding
Gene Aliases
CELE_F07C6.1
Chromosome Location
Chr.IV: 12793816 - 12796164 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268743.2 NP_001255672.1 1 - 2 166 70

View Full Product Details