Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dli-1
Entrez Gene ID
178260
Gene Name
protein-coding
Gene Aliases
CELE_C39E9.14
Chromosome Location
Chr.IV: 13096958 - 13099702 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070118.7 NP_502519.2 4 - 5 780 71

View Full Product Details