Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gcl-1
Entrez Gene ID
190457
Gene Name
BTB domain-containing protein
Gene Aliases
CELE_Y62E10A.16
Chromosome Location
Chr.IV: 13408325 - 13412895 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070184.5 NP_502585.1 1 - 2 206 78

View Full Product Details