Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pik-1
Entrez Gene ID
178306
Gene Name
protein-coding
Gene Aliases
CELE_K09B11.1
Chromosome Location
Chr.IV: 13416041 - 13422391 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268813.4 NP_001255742.1 2 - 3 189 83
NR_056884.1 2 - 3 217 83
GenBank mRNA AF348167.1 - 1 - 2 133 83

View Full Product Details