Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pik-1
Entrez Gene ID
178306
Gene Name
protein-coding
Gene Aliases
CELE_K09B11.1
Chromosome Location
Chr.IV: 13416041 - 13422391 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268813.4 NP_001255742.1 3 - 4 384 129
NM_001268814.3 NP_001255743.1 1 - 2 181 129
NR_056884.1 3 - 4 412 129
GenBank mRNA AF348167.1 - 2 - 3 328 129

View Full Product Details