Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
seld-1
Entrez Gene ID
189912
Gene Name
Probable selenide, water dikinase
Gene Aliases
CELE_Y45F10A.4
Chromosome Location
Chr.IV: 13503238 - 13509215 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383238.1 4 - 5 1102 96

View Full Product Details