Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tlp-1
Entrez Gene ID
178338
Gene Name
C2H2-type domain-containing protein
Gene Aliases
CELE_T23G4.1
Chromosome Location
Chr.IV: 13701208 - 13704082 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070246.5 NP_502647.1 3 - 4 942 92
GenBank mRNA AY046083.1 - 3 - 4 840 92

View Full Product Details