Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gei-18
Entrez Gene ID
178357
Gene Name
GEX Interacting protein
Gene Aliases
CELE_Y37A1B.15
Chromosome Location
Chr.IV: 13981062 - 13982525 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070280.5 NP_502681.3 2 - 3 172 169

View Full Product Details