Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
noah-2
Entrez Gene ID
178365
Gene Name
NOmpA Homolog (Drosophila nompA: no mechanoreceptor potential A)
Gene Aliases
CELE_F52B11.3
Chromosome Location
Chr.IV: 14091254 - 14097391 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070298.6 NP_502699.1 2 - 3 909 64

View Full Product Details