Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tra-3
Entrez Gene ID
178385
Gene Name
Calpain-5
Gene Aliases
CELE_LLC1.1
Chromosome Location
Chr.IV: 14436367 - 14464006 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070350.6 NP_502751.1 5 - 6 928 80
NR_131640.1 4 - 5 814 80

View Full Product Details