Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-63
Entrez Gene ID
3565760
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_T27E7.8
Chromosome Location
Chr.IV: 14523868 - 14525197 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028233.1 NP_001023404.1 1 - 2 351 135

View Full Product Details