Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rom-4
Entrez Gene ID
3565076
Gene Name
protein-coding
Gene Aliases
CELE_Y116A8C.16
Chromosome Location
Chr.IV: 16995822 - 17014953 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047548.5 NP_001041013.2 13 - 14 3437 76
NM_001083250.6 NP_001076719.2 13 - 14 3443 76
NM_001276901.3 NP_001263830.1 1 - 2 176 76
NM_001380564.1 NP_001368544.1 6 - 7 2492 76
NM_001380565.1 NP_001368545.1 4 - 5 1646 76

View Full Product Details