Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-15
Entrez Gene ID
187983
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_T02D1.3
Chromosome Location
Chr.IV: 17396210 - 17397491 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070702.1 NP_503103.1 3 - 4 603 125

View Full Product Details