Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pmp-4
Entrez Gene ID
24104835
Gene Name
Peroxisomal Membrane Protein related
Gene Aliases
CELE_T02D1.5
Chromosome Location
Chr.IV: 17402453 - 17405630 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306751.2 NP_001293680.1 1 - 2 296 151

View Full Product Details