Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-27
Entrez Gene ID
191654
Gene Name
protein-coding
Gene Aliases
CELE_C06A12.4
Chromosome Location
Chr.IV: 17432977 - 17436652 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269027.3 NP_001255956.2 1 - 2 87 157
NM_001269028.2 NP_001255957.2 1 - 2 66 157

View Full Product Details