Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-48
Entrez Gene ID
182756
Gene Name
One cut domain family member
Gene Aliases
CELE_C17H12.9
Chromosome Location
Chr.IV: 6812108 - 6815609 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068645.6 NP_501046.1 3 - 4 611 110

View Full Product Details