Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atg-9
Entrez Gene ID
178561
Gene Name
protein-coding
Gene Aliases
CELE_T22H9.2
Chromosome Location
Chr.V: 349385 - 359658 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070777.5 NP_503178.2 9 - 10 2319 57

View Full Product Details