Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-37
Entrez Gene ID
191856
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_R11G11.9
Chromosome Location
Chr.V: 511194 - 512520 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070827.1 NP_503228.1 2 - 3 322 136

View Full Product Details