Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-59
Entrez Gene ID
187745
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_R09E12.1
Chromosome Location
Chr.V: 778639 - 781888 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070923.3 NP_503324.2 3 - 4 631 66

View Full Product Details