Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snt-5
Entrez Gene ID
178631
Gene Name
protein-coding
Gene Aliases
CELE_R12A1.2
Chromosome Location
Chr.V: 1393112 - 1411559 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313245.4 NP_001300174.1 5 - 6 802 120
NM_001313246.3 NP_001300175.1 5 - 6 712 120
NM_001313247.3 NP_001300176.1 7 - 8 907 120
NM_071008.5 NP_503409.1 7 - 8 1003 120

View Full Product Details