Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-8
Entrez Gene ID
186126
Gene Name
Serpentine Receptor, class SX
Gene Aliases
CELE_F52F10.5
Chromosome Location
Chr.V: 1551008 - 1553104 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071043.5 NP_503444.4 4 - 5 724 145
NR_132502.1 4 - 5 640 145

View Full Product Details