Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-24
Entrez Gene ID
183491
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_C45H4.10
Chromosome Location
Chr.V: 2151758 - 2153092 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071181.2 NP_503582.1 4 - 5 635 141

View Full Product Details