Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sknr-1
Entrez Gene ID
189139
Gene Name
protein-coding
Gene Aliases
CELE_W02H5.7
Chromosome Location
Chr.V: 2622056 - 2625460 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316826.3 NP_001303755.1 1 - 2 188 148

View Full Product Details