Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kel-8
Entrez Gene ID
3565011
Gene Name
protein-coding
Gene Aliases
CELE_W02G9.2
Chromosome Location
Chr.V: 2671432 - 2688520 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313280.3 NP_001300209.1 6 - 6 1994 65
NM_001313281.3 NP_001300210.1 1 - 2 1307 65
NM_071328.8 NP_503729.4 6 - 7 2026 65
GenBank mRNA DQ338465.1 - 5 - 6 1883 65

View Full Product Details