Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srh-36
Entrez Gene ID
191855
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_T03D3.4
Chromosome Location
Chr.V: 2819462 - 2822248 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071363.2 NP_503764.1 3 - 4 726 84

View Full Product Details