Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-49
Entrez Gene ID
191950
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_T03D3.12
Chromosome Location
Chr.V: 2846250 - 2848340 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071369.1 NP_503770.1 2 - 3 558 108

View Full Product Details