Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-54
Entrez Gene ID
191953
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_F37B4.11
Chromosome Location
Chr.V: 2858660 - 2859838 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071373.1 NP_503774.1 3 - 4 661 123

View Full Product Details