Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-38
Entrez Gene ID
191857
Gene Name
protein-coding
Gene Aliases
CELE_C31B8.11
Chromosome Location
Chr.V: 2920600 - 2922694 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071392.1 NP_503793.1 2 - 3 324 91

View Full Product Details