Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-6
Entrez Gene ID
183259
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_C36C5.3
Chromosome Location
Chr.V: 3170138 - 3171408 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071473.2 NP_503874.2 1 - 2 123 111

View Full Product Details