Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scrm-3
Entrez Gene ID
182212
Gene Name
Phospholipid scramblase
Gene Aliases
CELE_C04E12.7
Chromosome Location
Chr.V: 3355848 - 3357209 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071533.3 NP_503934.2 2 - 3 583 68
GenBank mRNA EF445111.1 - 2 - 3 567 68

View Full Product Details