Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gst-33
Entrez Gene ID
178771
Gene Name
Glutathione S-Transferase
Gene Aliases
CELE_C02A12.1
Chromosome Location
Chr.V: 3467451 - 3468562 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071567.4 NP_503968.1 2 - 3 529 104

View Full Product Details