Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-35B1
Entrez Gene ID
178803
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_K07C6.4
Chromosome Location
Chr.V: 3939032 - 3941237 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322546.2 NP_001309511.1 2 - 3 416 67

View Full Product Details