Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-35A4
Entrez Gene ID
183622
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_C49G7.8
Chromosome Location
Chr.V: 4031619 - 4033446 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071723.2 NP_504124.1 3 - 4 471 134

View Full Product Details