Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nlp-13
Entrez Gene ID
178826
Gene Name
Neuropeptide-Like Protein
Gene Aliases
CELE_E03D2.1
Chromosome Location
Chr.V: 4236418 - 4238056 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071769.5 NP_504170.1 1 - 2 74 67

View Full Product Details