Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-12
Entrez Gene ID
184010
Gene Name
protein-coding
Gene Aliases
CELE_E03D2.4
Chromosome Location
Chr.V: 4240952 - 4242531 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071771.2 NP_504172.2 2 - 3 576 150

View Full Product Details