Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mcm-7
Entrez Gene ID
178831
Gene Name
DNA replication licensing factor MCM7
Gene Aliases
CELE_F32D1.10
Chromosome Location
Chr.V: 4347160 - 4349768 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071798.8 NP_504199.1 2 - 3 975 67

View Full Product Details